RNA collection tubes, BD PAXgene™
Supplier: BD Medical
762165
BDAM762165EA 5850 GBP
BDAM762165
RNA collection tubes, BD PAXgene™
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The tube contains a proprietary reagent that immediately stabilises all total RNA, including miRNA.
  • Enables accurate detection and quantitation of gene transcripts
  • Minimises ex vivo blood RNA degradation or gene induction
Stabilises RNA for three days at room temperature (18 to 25 °C) and five days at 4 °C. Paxgene™ tubes can be frozen at –20 °C or –70 °C for long term storage.