Benzoyl peroxide blend with 50% dicyclohexyl phthalate, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: Dibenzoyl peroxide

Stabilisation: 50% DiCyclohexyl phthalate
MQ200

B5907-1VL
B5907-1VLEA 27.5 GBP
B5907-1VL
Benzoyl peroxide blend with 50% dicyclohexyl phthalate, Sigma-Aldrich®
Benzoyl peroxide

Suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.


Formulated specifically for use with Histocryl and LR white acrylic resins.

Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
Melting Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003071
CAS Number: 94-36-0
EINECS: 202-327-6
UN: 3104
ADR: 5.2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34eca7fc3f742783ab8198b7071acb6732625c2b&vid=d7f5bb43713db565face74bec8fecc3e2f24d99c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) White (May contain off-coloured particles)
Appearance (Form) Powder
Infrared spectrum Conforms to Structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR