4-Iodobenzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: p-Iodobenzonitrile

B25104.06 B25104.14 B25104.22
B25104.06EA 59.5 GBP
B25104.06 B25104.14 B25104.22
4-Iodobenzonitrile ≥98%
4-Iodobenzonitrile

Purity: 98% CAS No.: 3058-39-7 Molecular Formula: C7H4IN Formula Weight: 229.02 Melting Point: 125-127° UN No.: 3439 MDL No.: MFCD00051310

Formula: C₇H₄IN
MW: 229.02 g/mol
Boiling Pt: 285
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00051310
CAS Number: 3058-39-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR