4-(Trifluoromethoxy)phenylboronic acid 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Dihydroxy(4-(trifluoromethoxy)phenyl)borane, (4-(Trifluoromethoxy)phenyl)metaboric acid, 4-(Trifluoromethoxy)benzeneboronic acid, 4-Trifluoromethoxyphenylboronic acid

B23233.14 B23233.03 B23233.06
B23233.14EA 0 GBP
B23233.14 B23233.03 B23233.06
4-(Trifluoromethoxy)phenylboronic acid 98
4-(Trifluoromethoxy)phenylboronic acid

Purity: 98% CAS No.: 139301-27-2 Molecular Formula: C7H6BF3O3 Formula Weight: 205.93 Melting Point: ca 120-125° Beilstein Reference: 8548201

Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: ∼ 120…125 °C
Storage Temperature: Ambient
MDL Number: MFCD01074648
CAS Number: 139301-27-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR