1-Ethynylcyclopentanol 98

Supplier: Thermo Fisher Scientific
Warning

B23109.06 B23109.14
B23109.06EA 63.7 GBP
B23109.06 B23109.14
1-Ethynylcyclopentanol 98
1-Ethynylcyclopentanol

Purity: 98+% CAS No.: 17356-19-3 Molecular Formula: C7H10O Formula Weight: 110.16 Flash Point: 48°(118°F) Refractive Index: 1.4740 Odour: Irritating Melting Point: 23-27° Boiling Point: 156-159° Beilstein Reference: 1924167 UN No.: 1325 MDL No.: MFCD00001365

Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 156…159 °C
Melting Pt: 23…27 °C
Density: 0.962
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003858
CAS Number: 17356-19-3
EINECS: 241-385-7
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR