(4-Bromobutyl)triphenylphosphonium bromide ≥98%

Supplier: Thermo Fisher Scientific
Warning

B23040.09 B23040.18
B23040.09EA 35.7 GBP
B23040.09 B23040.18
(4-Bromobutyl)triphenylphosphonium bromide ≥98%
(4-Bromobutyl)triphenylphosphonium bromide

Purity: 98% CAS No.: 7333-63-3 Molecular Formula: Br(CH2)4P(C6H5)3Br Formula Weight: 478.22 Melting Point: 206-212° Beilstein Reference: 3812291

Formula: C₂₂H₂₃Br₂P
MW: 478.21 g/mol
Melting Pt: 206…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00031636
CAS Number: 7333-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR