Immersion oil
Supplier: PanReac AppliChem
A0699.0100
APPCA0699.0100EA 45.7 GBP
APPCA0699.0100
Immersion oil
Stains Microscopy and staining reagents
This fluorescence-free immersion oil for microscopy is highly UV-permeable and free of PCB's.
Order Now
SPECIFICATIONS
Environmentally Preferable