Acetone 99,5 GC_ASSAY_METHOD, Liquid USP, Ph. Eur., BP, pure

Supplier: PanReac AppliChem
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

141007.1611 141007.1214 141007.0716 141007.0816 141007.1211 141007.1212
APPC141007.1611EA 41.7 GBP
APPC141007.1611 APPC141007.1214 APPC141007.0716 APPC141007.0816 APPC141007.1211 APPC141007.1212
Acetone 99,5 GC_ASSAY_METHOD, Liquid USP, Ph. Eur., BP, pure
Acetone

Synonyms: 2-Propanone, ß-Ketopropane, Dimethylketone, Pyroacetic Ether.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.5 °C
Melting Pt: -95.4
Density: 0.789 Kg/l
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility Water
Refractive index 20/D 1,3591
Appearance Liquid
Min. assay (G.C.) 99,5%
Identity according to pharmacopoeias Passes test
Density 25/25 ≤0,789
Density 20/20 0,790 - 0,793
Appearance of solution Passes test
Acidity Passes test
Alkalinity Passes test
Insoluble matter in H₂O Passes test
Non-volatile matter 0,004%
Reducing substance to KMnO₄ Passes test
Residual solvents (Ph. Eur./USP) Passes test
Methanol 0,05%
2-Propanol 0,05%
Benzene 0,0002%
Other impurities 0,05%
Ethanol (G.C.) 500 ppm
Water (H₂O) 0,3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR