Pentafluorophenylhydrazine 97%

Supplier: Apollo Scientific
Warning

PC5690-25G PC5690-5G PC5690-100G
APOSPC5690-25GEA 170 GBP
APOSPC5690-25G APOSPC5690-5G APOSPC5690-100G
Pentafluorophenylhydrazine 97%
Pentafluorophenylhydrazine
Formula: C₆H₃F₅N₂
MW: 198.1 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00007574
CAS Number: 828-73-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR