2,4,6-Tris[bromo(fluoro)methyl]-1,3,5-triazine 95%

Supplier: Apollo Scientific

PC51110-1G
APOSPC51110-1GEA 268 GBP
APOSPC51110-1G
2,4,6-Tris[bromo(fluoro)methyl]-1,3,5-triazine 95%
2,4,6-Tris[bromo(fluoro)methyl]-1,3,5-triazine
Formula: C₆H₃Br₃F₃N₃
MW: 413.815 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442115
CAS Number: 402-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR