DL-2-[4-(Trifluoromethoxy)phenoxy]propionic acid 97%

Supplier: Apollo Scientific
Warning

PC5044-1G
APOSPC5044-1GEA 198 GBP
APOSPC5044-1G
DL-2-[4-(Trifluoromethoxy)phenoxy]propionic acid 97%
DL-2-[4-(Trifluoromethoxy)phenoxy]propionic acid
Formula: C₁₀H₉F₃O₄
MW: 250.18 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00052353
CAS Number: 175204-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR