(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

PC4557-1G
APOSPC4557-1GEA 19.7 GBP
APOSPC4557-1G
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00236304
CAS Number: 239135-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR