3-(2,2-Difluoroethoxy)propionitrile

Supplier: Apollo Scientific

PC450254-1G
APOSPC450254-1GEA 388 GBP
APOSPC450254-1G
3-(2,2-Difluoroethoxy)propionitrile
3-(2,2-Difluoroethoxy)propionitrile
Formula: C₅H₇F₂NO
MW: 135.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442116
CAS Number: 1309602-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR