5-Bromo-2-(trifluoromethoxy)benzoic acid 99%

Supplier: Apollo Scientific
Warning

PC408237-1G PC408237-25G PC408237-5G
APOSPC408237-1GEA 39.3 GBP
APOSPC408237-1G APOSPC408237-25G APOSPC408237-5G
5-Bromo-2-(trifluoromethoxy)benzoic acid 99%
5-Bromo-2-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203214
CAS Number: 403646-47-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR