3-[4-Methoxy-3-(trifluoromethoxy)phenyl]propionic acid 97%

Supplier: Apollo Scientific

Synonyms: 3-(4-Methoxy-3-(trifluoromethoxy)phenyl)propanoic acid

PC302825-1G
APOSPC302825-1GEA 208 GBP
APOSPC302825-1G
3-[4-Methoxy-3-(trifluoromethoxy)phenyl]propionic acid 97%
3-[4-Methoxy-3-(trifluoromethoxy)phenyl]propionic acid
Formula: C₁₁H₁₁F₃O₄
MW: 264.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398561
CAS Number: 1261587-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR