2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile

Supplier: Apollo Scientific
Warning

PC200202-1G
APOSPC200202-1GEA 119 GBP
APOSPC200202-1G
2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
Formula: C₈H₄ClF₃N₂
MW: 220.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568352
CAS Number: 13600-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR