3-(Trifluoromethoxy)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: (3-(Trifluoromethoxy)phenyl)metaboric acid, (3-(Trifluoromethoxy)phenyl)boronic acid, 3-(Trifluoromethoxy)benzeneboronic acid, Dihydroxy(3-(trifluoromethoxy)phenyl)borane

PC0240-25G PC0240-5G
APOSPC0240-25GEA 78.5 GBP
APOSPC0240-25G APOSPC0240-5G
3-(Trifluoromethoxy)phenylboronic acid 98%
3-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: 80…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320697
CAS Number: 179113-90-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR