p-Toluenesulphonyl chloride 97%

Supplier: Apollo Scientific
Danger

Synonyms: Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-p-sulphonyl chloride

OR9931-100G OR9931-1KG
APOSOR9931-100GEA 19.7 GBP
APOSOR9931-100G APOSOR9931-1KG
p-Toluenesulphonyl chloride 97%
p-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 146 °C/15mmHg
Melting Pt: 67…70 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007450
CAS Number: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8 II
Merck Index: 13,09612

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR