1-(4-Pyridyl)-1-butylamine 95%

Supplier: Apollo Scientific

Synonyms: 1-(Pyridin-4-yl)butan-1-amine

OR964746-1G
APOSOR964746-1GEA 335 GBP
APOSOR964746-1G
1-(4-Pyridyl)-1-butylamine 95%
1-(4-Pyridyl)-1-butylamine
Formula: C₉H₁₄N₂
MW: 150.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153946
CAS Number: 1179877-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR