2-Chloronaphthalene 96%

Supplier: Apollo Scientific
Danger

OR963893-1G OR963893-5G OR963893-25G
APOSOR963893-1GEA 19.7 GBP
APOSOR963893-1G APOSOR963893-5G APOSOR963893-25G
2-Chloronaphthalene 96%
2-Chloronaphthalene
Formula: C₁₀H₇Cl
MW: 162.62 g/mol
Boiling Pt: 256 °C
Melting Pt: 57…61 °C
MDL Number: MFCD00035731
CAS Number: 91-58-7
Merck Index: 13,02169

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR