6-Chloro-2-(dimethylamino)nicotinaldehyde 98%

Supplier: Apollo Scientific

OR959963-1G OR959963-5G
APOSOR959963-1GEA 262 GBP
APOSOR959963-1G APOSOR959963-5G
6-Chloro-2-(dimethylamino)nicotinaldehyde 98%
6-Chloro-2-(dimethylamino)nicotinaldehyde
Formula: C₈H₉ClN₂O
MW: 184.623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21333091
CAS Number: 1233698-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR