2,1,3-Benzoxadiazole-4-carbaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 4-Benzofurazancarboxaldehyde, 4-Formylbenzofurazan, 2,1,3-Benzoxadiazole-4-carboxaldehyde

OR951625-5G
APOSOR951625-5GEA 118 GBP
APOSOR951625-5G
2,1,3-Benzoxadiazole-4-carbaldehyde
2,1,3-Benzoxadiazole-4-carbaldehyde
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Melting Pt: 108…109 °C
Storage Temperature: Ambient
MDL Number: MFCD07772900
CAS Number: 32863-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR