Allyl cinnamate 98%

Supplier: Apollo Scientific

Synonyms: Allyl Cinnamate, Cinnamic Acid Allyl Ester, Cinnamic acid Allyl Ester

OR919186-100G OR919186-1KG OR919186-500G
APOSOR919186-100GEA 112 GBP
APOSOR919186-100G APOSOR919186-1KG APOSOR919186-500G
Allyl cinnamate 98%
Allyl cinnamate
Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026105
CAS Number: 1866-31-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR