4-[2-(Dimethylamino)ethyl]aniline 97%

Supplier: Apollo Scientific

OR918366-10G OR918366-1G OR918366-5G
APOSOR918366-10GEA 273 GBP
APOSOR918366-10G APOSOR918366-1G APOSOR918366-5G
4-[2-(Dimethylamino)ethyl]aniline 97%
4-[2-(Dimethylamino)ethyl]aniline
Formula: C₁₀H₁₆N₂
MW: 164.247 g/mol
Boiling Pt: 137…138 °C (6 mm Hg)
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD04114509
CAS Number: 5636-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR