1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 97%

Supplier: Apollo Scientific

Synonyms: Cyclohexane-1,3-dicarboxylic acid, 1,3-Cyclohexanedicarboxylic acid

OR917608-100G
APOSOR917608-100GEA 101 GBP
APOSOR917608-100G
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 97%
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134411
CAS Number: 3971-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR