tert-Butyldimethylsilyl-(S)-glycidyl ether 95%

Supplier: Apollo Scientific

Synonyms: tert-Butyldimethylsilyl (S)-(+)-glycidyl ether, (S)-1-[(tert-Butyldimethylsilyl)oxy]-2,3-epoxypropane, (S)-2-[(tert-Butyldimethylsilyloxy)methyl]oxirane

OR917497-5G
APOSOR917497-5GEA 56.3 GBP
APOSOR917497-5G
tert-Butyldimethylsilyl-(S)-glycidyl ether 95%
tert-Butyldimethylsilyl-(S)-glycidyl ether
Formula: C₉H₂₀O₂Si
MW: 188.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862148
CAS Number: 123237-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR