trans-4-Hydroxy-L(-)-proline

Supplier: Apollo Scientific
Warning

Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH

OR6733-100G OR6733-500G OR6733-25G
APOSOR6733-100GEA 19.7 GBP
APOSOR6733-100G APOSOR6733-500G APOSOR6733-25G
trans-4-Hydroxy-L(-)-proline
trans-4-Hydroxy-L(-)-proline
Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 296
Melting Pt: 270…273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR