Indole-4-carbonitrile

Supplier: Apollo Scientific
Danger

Synonyms: 4-Cyano-1H-indole, 1H-Indole-4-carbonitrile, 4-Cyanoindole

OR59809-5G
APOSOR59809-5GEA 19.7 GBP
APOSOR59809-5G
Indole-4-carbonitrile
Indole-4-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00152045
CAS Number: 16136-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR