2,4,6-Collidine

Supplier: Apollo Scientific
Danger

OR4784-100ML OR4784-500ML
APOSOR4784-100MLEA 78.5 GBP
APOSOR4784-100ML APOSOR4784-500ML
2,4,6-Collidine
2,4,6-Collidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 171…172 °C
Melting Pt: –43 °C
Density: 0.917 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006338
CAS Number: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3+6.1 III
Merck Index: 13,09789

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR