4-Bromo-4'-(diphenylamino)biphenyl

Supplier: Apollo Scientific

Synonyms: 4'-Bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine, (4'-Bromobiphenyl-4-yl)diphenylamine

OR471453-5G
APOSOR471453-5GEA 75.9 GBP
APOSOR471453-5G
4-Bromo-4'-(diphenylamino)biphenyl
4-Bromo-4'-(diphenylamino)biphenyl
Formula: C₂₄H₁₈BrN
MW: 400.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038499
CAS Number: 202831-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR