2',4'-Dibromoacetophenone

Supplier: Apollo Scientific

OR471302-1G
APOSOR471302-1GEA 57.6 GBP
APOSOR471302-1G
2',4'-Dibromoacetophenone
2',4'-Dibromoacetophenone
Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152057
CAS Number: 33243-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR