Methyl 2-(4-Vinylphenyl)acetate

Supplier: Apollo Scientific

OR471170-1G
APOSOR471170-1GEA 726 GBP
APOSOR471170-1G
Methyl 2-(4-Vinylphenyl)acetate
Methyl 2-(4-Vinylphenyl)acetate
Formula: C₁₁H₁₂O₂
MW: 176.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18431776
CAS Number: 62667-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR