Methyl (R)-2-(3-Piperidyl)acetate hydrochloride 95%

Supplier: Apollo Scientific
Warning

OR471028-1G
APOSOR471028-1GEA 753 GBP
APOSOR471028-1G
Methyl (R)-2-(3-Piperidyl)acetate hydrochloride 95%
Methyl (R)-2-(3-Piperidyl)acetate hydrochloride
Formula: C₈H₁₅NO₂·ClH
MW: 193.671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094723
CAS Number: 865157-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR