3-Cyclopropyl-2-propenoic acid ethyl ester

Supplier: Apollo Scientific

OR451080-1G
APOSOR451080-1GEA 213 GBP
APOSOR451080-1G
3-Cyclopropyl-2-propenoic acid ethyl ester
3-Cyclopropyl-2-propenoic acid ethyl ester
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11506679
CAS Number: 5808-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR