(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid 95+%

Supplier: Apollo Scientific

Synonyms: (S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid, (2S)-3-Amino-2-benzylpropanoic acid, N-BOC protected, (2S)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-phenylpropanoic acid

OR40213-250MG
APOSOR40213-250MGEA 354 GBP
APOSOR40213-250MG
(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid 95+%
(S)-2-Benzyl-3-((tert-butoxycarbonyl)amino)propanoic acid
Formula: C₁₅H₂₁NO₄
MW: 279.332 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07372495
CAS Number: 189619-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR