3-(Morpholino)phenylboronic acid 97%

Supplier: Apollo Scientific

Synonyms: (3-Morpholinophenyl)boronic acid

OR360139-5G
APOSOR360139-5GEA 161 GBP
APOSOR360139-5G
3-(Morpholino)phenylboronic acid 97%
3-(Morpholino)phenylboronic acid
Formula: C₁₀H₁₄BNO₃
MW: 207.0341 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412095
CAS Number: 863377-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR