Methyl trans-cinnamate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl (E)-cinnamate, trans-Cinnamic acid methyl ester

OR350234-100G OR350234-500G
APOSOR350234-100GEA 23.6 GBP
APOSOR350234-100G APOSOR350234-500G
Methyl trans-cinnamate 95%
Methyl trans-cinnamate
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 262 °C (760 mm Hg)
Melting Pt: 34 °C
Density: 1.08
Storage Temperature: Refrigerator
MDL Number: MFCD00008458
CAS Number: 1754-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR