5-Amino-3-methyl-1-p-tolylpyrazole 95%

Supplier: Apollo Scientific

Synonyms: 3-Methyl-1-(4-methylphenyl)-1H-pyrazol-5-amine

OR32925-5G
APOSOR32925-5GEA 87.7 GBP
APOSOR32925-5G
5-Amino-3-methyl-1-p-tolylpyrazole 95%
5-Amino-3-methyl-1-p-tolylpyrazole
Formula: C₁₁H₁₃N₃
MW: 187.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020725
CAS Number: 62535-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR