1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde 95%

Supplier: Apollo Scientific

OR32718-1G
APOSOR32718-1GEA 485 GBP
APOSOR32718-1G
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde 95%
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde
Formula: C₉H₆ClN₃O
MW: 207.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01109282
CAS Number: 113934-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR