Isochroman-1-ylmethanamine 95%

Supplier: Apollo Scientific
Danger

Synonyms: (3,4-Dihydro-1H-isochromen-1-ylmethyl)amine, C-Isochroman-1-ylmethylamine

OR322556-100MG OR322556-250MG OR322556-50MG
APOSOR322556-100MGEA 105 GBP
APOSOR322556-100MG APOSOR322556-250MG APOSOR322556-50MG
Isochroman-1-ylmethanamine 95%
Isochroman-1-ylmethanamine
Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
MDL Number: MFCD00457645
CAS Number: 19158-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR