Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride

Supplier: Apollo Scientific

Synonyms: Z-Dap-OMe·HCl, Methyl 2-(S)-[N-Carbobenzyloxy]amino-3-aminopropionate, Hydrochloride

OR322488-250MG
APOSOR322488-250MGEA 30.2 GBP
APOSOR322488-250MG
Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride
Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride
Formula: C₁₂H₁₆N₂O₄·ClH
MW: 288.727 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701202
CAS Number: 35761-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR