1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

Supplier: Apollo Scientific

OR310731-1G
APOSOR310731-1GEA 704 GBP
APOSOR310731-1G
1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
Formula: C₁₃H₁₈Cl₃N₅
MW: 350.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28023519
CAS Number: 1610377-07-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR