(2-Iodo-5-methoxyphenyl)boronic acid

Supplier: Apollo Scientific

OR310361-1G
APOSOR310361-1GEA 30.2 GBP
APOSOR310361-1G
(2-Iodo-5-methoxyphenyl)boronic acid
(2-Iodo-5-methoxyphenyl)boronic acid
Formula: C₇H₈BIO₃
MW: 277.852 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26793796
CAS Number: 89694-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR