Cyclobutanone 98%

Supplier: Apollo Scientific
Danger

OR30488-25G OR30488-5G OR30488-100G
APOSOR30488-25GEA 44.5 GBP
APOSOR30488-25G APOSOR30488-5G APOSOR30488-100G
Cyclobutanone 98%
Cyclobutanone
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 97…99 °C
Melting Pt: –51 °C
Density: 0.938 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001332
CAS Number: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR