Methyl (3-methoxyphenyl)acetate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3-Methoxyphenylacetate

OR21784-1G OR21784-25G OR21784-5G
APOSOR21784-1GEA 19.7 GBP
APOSOR21784-1G APOSOR21784-25G APOSOR21784-5G
Methyl (3-methoxyphenyl)acetate 98%
Methyl (3-methoxyphenyl)acetate
Formula: C₁₀H₁₂O₃
MW: 180.23 g/mol
Boiling Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00017205
CAS Number: 18927-05-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR