3-Phenylpyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3-Phenylpicolinic acid

OR16992-1G
APOSOR16992-1GEA 250 GBP
APOSOR16992-1G
3-Phenylpyridine-2-carboxylic acid
3-Phenylpyridine-2-carboxylic acid
Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114112
CAS Number: 103863-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR