Methyl-3-methoxy-4-nitrobenzoate

Supplier: Apollo Scientific

Synonyms: Methyl 3-methoxy-4-nitrobenzoate, 3-Methoxy-4-nitrobenzoic acid methyl ester, Methyl 3-methoxy-4-nitrobenzenecarboxylate

OR14144-10G OR14144-1G OR14144-5G
APOSOR14144-10GEA 172 GBP
APOSOR14144-10G APOSOR14144-1G APOSOR14144-5G
Methyl-3-methoxy-4-nitrobenzoate
Methyl 3-methoxy-4-nitrobenzoate
Formula: C₉H₉NO₅
MW: 211.17 g/mol
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00051967
CAS Number: 5081-37-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR