1-(2,4-Dichloro-3,5-dimethylphenoxy)acetone

Supplier: Apollo Scientific

OR111604-1G
APOSOR111604-1GEA 280 GBP
APOSOR111604-1G
1-(2,4-Dichloro-3,5-dimethylphenoxy)acetone
1-(2,4-Dichloro-3,5-dimethylphenoxy)acetone
Formula: C₁₁H₁₂Cl₂O₂
MW: 247.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32069498
CAS Number: 2415751-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR