Diethyl (4-carboxybut-1-yl)phosphonate

Supplier: Apollo Scientific
Warning

Synonyms: 5-(Diethoxyphosphoryl)pentanoic acid, 5-(Diethylphosphono)pentanoic acid

OR10733-1G
APOSOR10733-1GEA 189 GBP
APOSOR10733-1G
Diethyl (4-carboxybut-1-yl)phosphonate
Diethyl (4-carboxybut-1-yl)phosphonate
Formula: C₉H₁₉O₅P
MW: 238.218 g/mol
Melting Pt: 19…20 °C
Density: 1.1401 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00800260
CAS Number: 55881-52-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR