3-Methylbenzamidoxime

Supplier: Apollo Scientific
Warning

Synonyms: N-Hydroxy-3-methylbenzimidamide

OR0604-1G OR0604-5G
APOSOR0604-1GEA 40.7 GBP
APOSOR0604-1G APOSOR0604-5G
3-Methylbenzamidoxime
3-Methylbenzamidoxime
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00833266
CAS Number: 40067-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR